Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027902.2 | NP_082178.2 | 3 | 539 | ||
GenBank mRNA | AK004939.2 | - | 3 | 386 | ||
AY240929.1 | - | 3 | 516 | |||
BC029645.1 | - | 3 | 516 | 4195486 | ||
BC057674.1 | - | 3 | 481 | 4162007 | ||
EU190436.1 | - | 2 | 330 | |||
EU190437.1 | - | 2 | 330 |