Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027902.2 | NP_082178.2 | 18 | 3011 | ||
GenBank mRNA | AK004939.2 | - | 18 | 2861 | ||
AY240929.1 | - | 18 | 2988 | |||
BC029645.1 | - | 18 | 2988 | 4195486 | ||
BC057674.1 | - | 18 | 2953 | 4162007 |