Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163729.2 | NP_001157201.2 | 3 | 605 | ||
NM_027910.3 | NP_082186.2 | 4 | 758 | |||
GenBank mRNA | AB053465.1 | - | 4 | 629 | ||
AJ278127.1 | - | 4 | 625 | |||
AK004964.1 | - | 4 | 735 | |||
AK141673.1 | - | 3 | 590 | |||
AK170623.1 | - | 4 | 630 | |||
BC018154.1 | - | 4 | 727 | 3983436 |