Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027927.3 | NP_082203.1 | 3 | 298 | ||
GenBank mRNA | AK003874.1 | - | 3 | 310 | ||
AK012829.1 | - | 3 | 311 | |||
AK038713.1 | - | 2 | 1804 | |||
AK155162.1 | - | 3 | 310 | |||
BC023801.1 | - | 3 | 298 | 5353058 | ||
BC046996.1 | - | 3 | 292 | 6494551 |