Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027927.3 | NP_082203.1 | 6 | 772 | ||
GenBank mRNA | AK003874.1 | - | 6 | 784 | ||
AK155162.1 | - | 6 | 784 | |||
BC010657.1 | - | 4 | 444 | |||
BC023801.1 | - | 6 | 772 | 5353058 | ||
BC046996.1 | - | 6 | 766 | 6494551 |