Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027950.1 | NP_082226.1 | 6 | 1605 | ||
GenBank mRNA | AK005892.1 | - | 4 | 1368 | ||
AK156249.1 | - | 1 | 2094 | |||
BC006032.1 | - | 5 | 1946 | |||
BC022135.1 | - | 6 | 1605 | 5099665 | ||
BC091765.1 | - | 1 | 813 | |||
BC110674.1 | - | 6 | 1590 | |||
BC132440.1 | - | 6 | 1574 | 40130717 | ||
BC138192.1 | - | 6 | 1574 | 8861211 |