Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168244.1 | NP_001161716.1 | 2, 3 | 737 | ||
NM_001168246.1 | NP_001161718.1 | 3, 4 | 436 | |||
NM_027954.3 | NP_082230.2 | 3, 4 | 418 | |||
GenBank mRNA | AK005954.1 | - | 3, 4 | 419 | ||
AK082833.1 | - | 2, 3 | 304 | |||
AK167332.1 | - | 2, 3 | 736 | |||
BC094628.1 | - | 3, 4 | 313 | 3374002 | ||
BC132534.1 | - | 3, 4 | 368 | 40130811 | ||
BY246600.1 | - | 3, 4 | 351 |