Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006402.2 | NP_006393.2 | 2, 3 | 394 | ||
GenBank mRNA | AF029890.1 | - | 2, 3 | 145 | ||
AK054769.1 | - | 1, 2 | 1370 | |||
AY623819.1 | - | 2, 3 | 90 | |||
BC062619.1 | - | 2, 3 | 412 | 5190406 | ||
BX092018.1 | - | 2, 3 | 410 | |||
CR456866.1 | - | 2, 3 | 90 | |||
CR542130.1 | - | 2, 3 | 90 |