Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310067B10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028014.3 | NP_082290.2 | 22 | 3130 | ||
GenBank mRNA | AK047622.1 | - | 8 | 1264 | ||
AK155233.1 | - | 22 | 3005 | |||
BC007157.1 | - | 9 | 1376 | |||
BC030865.1 | - | 7 | 897 | |||
BC053088.1 | - | 22 | 3045 | 5709104 | ||
BC115845.1 | - | 4 | 730 | |||
BC148215.1 | - | 3 | 807 |