Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028019.3 | NP_082295.1 | 4 | 756 | ||
GenBank mRNA | AK010429.1 | - | 4 | 758 | ||
AK150921.1 | - | 4 | 757 | |||
AK152096.1 | - | 4 | 749 | |||
AK185113.1 | - | 3 | 346 | |||
AK206997.1 | - | 3 | 346 | |||
BC096385.1 | - | 4 | 750 | 30744704 | ||
BC132307.1 | - | 4 | 752 | 40130584 | ||
BC138303.1 | - | 4 | 752 | 8861322 | ||
BC145236.1 | - | 3 | 588 |