Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028019.3 | NP_082295.1 | 5 | 1489 | ||
GenBank mRNA | AK002781.1 | - | 2 | 660 | ||
AK010429.1 | - | 5 | 1491 | |||
AK152096.1 | - | 5 | 1482 | |||
BC025209.1 | - | 1 | 652 | |||
BC096385.1 | - | 5 | 1483 | 30744704 |