Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127348.1 | NP_001120820.1 | 7 | 2343 | ||
NM_001127349.1 | NP_001120821.1 | 7 | 2348 | |||
NM_028035.4 | NP_082311.3 | 7 | 2446 | |||
GenBank mRNA | AK028163.1 | - | 7 | 2445 | ||
AK150704.1 | - | 7 | 2350 | |||
AK150878.1 | - | 7 | 2265 | |||
AK150999.1 | - | 7 | 2351 | |||
AK151320.1 | - | 7 | 2350 | |||
AK151520.1 | - | 7 | 2350 | |||
AK152825.1 | - | 7 | 2320 | |||
AK152949.1 | - | 7 | 2336 | |||
AK153127.1 | - | 7 | 2350 | |||
AK153356.1 | - | 7 | 2301 | |||
AK166121.1 | - | 7 | 2317 | |||
AK169110.1 | - | 7 | 2282 | |||
BC010334.1 | - | 7 | 2280 | 2655824 |