Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028039.2 | NP_082315.3 | 3 | 515 | ||
GenBank mRNA | AK050845.1 | - | 3 | 466 | ||
AK083728.1 | - | 1 | 294 | |||
AK144685.1 | - | 3 | 471 | |||
AK166457.1 | - | 3 | 463 | |||
BC033303.1 | - | 3 | 430 | 3675568 | ||
BC071275.1 | - | 3 | 448 | 6331551 | ||
CF168369.1 | - | 3 | 498 | |||
CF730483.1 | - | 1 | 35 |