Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028041.2 | NP_082317.1 | 6, 7 | 653 | ||
GenBank mRNA | AF319547.1 | - | 6, 7 | 1013 | ||
AK010484.1 | - | 3, 4 | 233 | |||
AK028274.1 | - | 6, 7 | 667 | |||
AK031534.1 | - | 6, 7 | 663 | |||
AK031679.1 | - | 6, 7 | 671 | |||
AK147715.1 | - | 6, 7 | 668 | |||
AK152989.1 | - | 6, 7 | 667 | |||
AK155138.1 | - | 6, 7 | 665 | |||
AK167603.1 | - | 6, 7 | 667 | |||
AK168455.1 | - | 6, 7 | 667 | |||
AK169255.1 | - | 6, 7 | 666 | |||
AK170967.1 | - | 6, 7 | 665 | |||
AK212480.1 | - | 2, 3 | 80 | |||
BC043699.1 | - | 6, 7 | 653 | 5344158 | ||
BC055026.1 | - | 2, 3 | 96 | |||
BC062920.1 | - | 6, 7 | 656 |