Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028044.2 | NP_082320.1 | 3 | 461 | ||
GenBank mRNA | AK005460.1 | - | 3 | 463 | ||
AK051121.1 | - | 2 | 679 | |||
AK051628.1 | - | 3 | 479 | |||
AK081672.1 | - | 3 | 377 | |||
AK159555.1 | - | 3 | 506 | |||
AK159871.1 | - | 3 | 491 | |||
AK166578.1 | - | 3 | 469 | |||
AK167201.1 | - | 3 | 482 | |||
AK167613.1 | - | 3 | 479 | |||
AK169021.1 | - | 3 | 486 | |||
AK169098.1 | - | 3 | 506 | |||
AK169410.1 | - | 3 | 436 | |||
AK179383.1 | - | 3 | 192 | |||
AK180041.1 | - | 2 | 211 | |||
AK190584.1 | - | 2 | 206 | |||
AK208457.1 | - | 2 | 161 | |||
AK212516.1 | - | 2 | 110 | |||
AK215736.1 | - | 2 | 171 | |||
AK216318.1 | - | 3 | 192 | |||
BC055711.1 | - | 3 | 462 | 6415164 | ||
BC085268.1 | - | 3 | 239 | 6487559 |