Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163032.1 | NP_001156504.1 | 5 | 1879 | ||
NM_028052.4 | NP_082328.3 | 6 | 1773 | |||
GenBank mRNA | AK005132.1 | - | 6 | 1694 | ||
AK032442.1 | - | 5 | 1781 | |||
AK049661.1 | - | 5 | 1725 | |||
AK078223.1 | - | 6 | 1774 | |||
BC026512.1 | - | 6 | 1720 | 4527827 |