Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104546.1 | NP_001098016.1 | 2, 3 | 181 | ||
NM_006413.4 | NP_006404.1 | 2, 3 | 181 | |||
XM_011539120.2 | XP_011537422.1 | 5, 6 | 734 | |||
XM_011539121.2 | XP_011537423.1 | 5, 6 | 734 | |||
XM_017015475.1 | XP_016870964.1 | 5, 6 | 734 | |||
XM_017015476.1 | XP_016870965.1 | 5, 6 | 734 | |||
XM_017015477.1 | XP_016870966.1 | 3, 4 | 434 | |||
XM_017015478.1 | XP_016870967.1 | 5, 6 | 734 | |||
GenBank mRNA | AK225532.1 | - | 2, 3 | 154 | ||
AK296196.1 | - | 2, 3 | 159 | |||
AK312900.1 | - | 2, 3 | 155 | |||
AL833628.1 | - | 2, 3 | 167 | |||
BC006991.1 | - | 3, 4 | 440 | 3827681 | ||
DB049454.1 | - | 2, 3 | 181 | |||
U77665.1 | - | 2, 3 | 173 |