Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028058.3 | NP_082334.1 | 4 | 470 | ||
GenBank mRNA | AK005163.1 | - | 4 | 471 | ||
AK007686.1 | - | 3 | 507 | |||
AK168868.1 | - | 4 | 457 | |||
AK210161.1 | - | 3 | 231 | |||
BC012515.1 | - | 4 | 412 | 2651125 | ||
BC030487.1 | - | 4 | 476 | 5374103 |