Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114322.1 | NP_001107794.1 | 8 | 928 | ||
NM_028069.3 | NP_082345.1 | 8 | 928 | |||
GenBank mRNA | AF462391.1 | - | 8 | 917 | ||
AK007988.1 | - | 8 | 917 | |||
AK027913.1 | - | 8 | 929 | |||
BC043336.1 | - | 8 | 893 | 5042289 | ||
BC054469.1 | - | 8 | 913 | 4225164 |