Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028074.4 | NP_082350.3 | 6 | 731 | ||
GenBank mRNA | AK049311.1 | - | 6 | 733 | ||
AK134182.1 | - | 5 | 510 | |||
AK169816.1 | - | 6 | 677 | |||
AK171730.1 | - | 6 | 600 | |||
AK209812.1 | - | 2 | 55 | |||
BC043036.1 | - | 5 | 488 | 5698965 |