Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028074.4 | NP_082350.3 | 8, 9 | 1102 | ||
GenBank mRNA | AK049311.1 | - | 8, 9 | 1104 | ||
AK134182.1 | - | 7, 8 | 908 | |||
AK169816.1 | - | 8, 9 | 1048 | |||
AK171730.1 | - | 8, 9 | 971 | |||
BC043036.1 | - | 7, 8 | 859 | 5698965 |