Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028108.3 | NP_082384.1 | 5 | 2412 | ||
GenBank mRNA | AI119854.1 | - | 1 | 221 | ||
AK135658.1 | - | 5 | 2355 | |||
AK145345.1 | - | 5 | 2340 | |||
AK167141.1 | - | 6 | 2532 | |||
BC057117.1 | - | 5 | 2372 | 6409354 |