Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Seh1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028112.2 | NP_082388.1 | 8 | 1699 | ||
GenBank mRNA | AK011330.1 | - | 8 | 1601 |