Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028136.2 | NP_082412.2 | 2, 3 | 396 | ||
GenBank mRNA | AF448804.1 | - | 2, 3 | 396 | ||
AK129373.1 | - | 2, 3 | 375 | |||
BC006615.1 | - | 2, 3 | 396 | |||
BC064890.1 | - | 2, 3 | 373 | |||
BC138061.1 | - | 2, 3 | 372 | 8861080 |