Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028140.4 | NP_082416.2 | 2 | 250 | ||
GenBank mRNA | AK013144.1 | - | 2 | 142 | ||
AK032080.1 | - | 2 | 271 | |||
AK035291.1 | - | 2 | 213 | |||
AK077957.1 | - | 2 | 251 | |||
BC010483.1 | - | 2 | 99 | |||
BC113183.1 | - | 2 | 181 | 40061891 | ||
BC113790.1 | - | 2 | 181 | 40061889 |