Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285991.1 | NP_001272920.1 | 4 | 479 | ||
NM_001285993.1 | NP_001272922.1 | 4 | 446 | |||
NM_028162.4 | NP_082438.3 | 4 | 479 | |||
GenBank mRNA | AK005461.1 | - | 4 | 463 | ||
AK033710.1 | - | 4 | 371 | |||
AK144467.1 | - | 4 | 480 | |||
AK154975.1 | - | 4 | 439 | |||
AK163366.1 | - | 3 | 288 | |||
AK220251.1 | - | 4 | 446 | |||
BC043113.1 | - | 4 | 295 | 6407234 | ||
BC098328.1 | - | 4 | 311 | 30357143 | ||
BY733740.1 | - | 4 | 476 |