Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028304.1 | - | 4 | 482 | ||
AK088814.1 | - | 4 | 473 | |||
AK142658.1 | - | 4 | 473 | |||
AK145291.1 | - | 4 | 468 | |||
AK146207.1 | - | 4 | 472 | |||
AK150551.1 | - | 4 | 479 | |||
AK150777.1 | - | 4 | 437 | |||
AK151240.1 | - | 4 | 436 | |||
AK151409.1 | - | 4 | 479 | |||
AK151664.1 | - | 4 | 479 | |||
AK152338.1 | - | 4 | 479 | |||
AK152343.1 | - | 4 | 438 | |||
AK152991.1 | - | 4 | 438 | |||
AK153379.1 | - | 4 | 479 | |||
AK155905.1 | - | 4 | 473 | |||
AK159203.1 | - | 4 | 473 | |||
AK159350.1 | - | 4 | 473 | |||
AK159878.1 | - | 4 | 482 | |||
AK164347.1 | - | 4 | 385 | |||
AK168708.1 | - | 4 | 415 | |||
AY029764.1 | - | 4 | 365 | |||
BC012401.1 | - | 4 | 462 | 3967323 | ||
CT010238.1 | - | 4 | 330 |