Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028304.1 | - | 4 | 505 | ||
AK088814.1 | - | 4 | 496 | |||
AK142658.1 | - | 4 | 496 | |||
AK145291.1 | - | 4 | 491 | |||
AK146207.1 | - | 4 | 495 | |||
AK150551.1 | - | 4 | 502 | |||
AK150777.1 | - | 4 | 460 | |||
AK151240.1 | - | 4 | 459 | |||
AK151409.1 | - | 4 | 502 | |||
AK151664.1 | - | 4 | 502 | |||
AK152338.1 | - | 4 | 502 | |||
AK152343.1 | - | 4 | 461 | |||
AK152991.1 | - | 4 | 461 | |||
AK153379.1 | - | 4 | 502 | |||
AK155905.1 | - | 4 | 496 | |||
AK159203.1 | - | 4 | 496 | |||
AK159350.1 | - | 4 | 496 | |||
AK159878.1 | - | 4 | 505 | |||
AK164347.1 | - | 4 | 408 | |||
AK168708.1 | - | 4 | 438 | |||
AY029764.1 | - | 4 | 388 | |||
BC012401.1 | - | 4 | 485 | 3967323 | ||
CT010238.1 | - | 4 | 353 |