Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007839.1 | - | 4 | 549 | ||
AK028304.1 | - | 11 | 1346 | |||
AK088814.1 | - | 11 | 1336 | |||
AK142658.1 | - | 11 | 1336 | |||
AK146207.1 | - | 11 | 1335 | |||
AK150551.1 | - | 11 | 1342 | |||
AK150777.1 | - | 11 | 1300 | |||
AK151240.1 | - | 11 | 1299 | |||
AK151409.1 | - | 11 | 1342 | |||
AK151664.1 | - | 11 | 1342 | |||
AK152338.1 | - | 11 | 1342 | |||
AK152343.1 | - | 11 | 1301 | |||
AK152991.1 | - | 11 | 1301 | |||
AK153379.1 | - | 11 | 1342 | |||
AK155905.1 | - | 11 | 1336 | |||
AK159203.1 | - | 11 | 1335 | |||
AK159350.1 | - | 11 | 1335 | |||
AK159878.1 | - | 11 | 1345 | |||
AK168708.1 | - | 11 | 1278 | |||
AY029764.1 | - | 11 | 1228 | |||
BC012401.1 | - | 11 | 1325 | 3967323 |