Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028186.4 | NP_082462.3 | 4 | 419 | ||
GenBank mRNA | AF358136.1 | - | 4 | 369 | ||
AK008814.1 | - | 4 | 401 | |||
AK081904.1 | - | 3 | 365 | |||
AK145155.1 | - | 4 | 401 | |||
AK146667.1 | - | 4 | 401 | |||
AK158350.1 | - | 4 | 401 | |||
AK165116.1 | - | 3 | 365 | |||
AK199881.1 | - | 2 | 81 | |||
AK203955.1 | - | 2 | 82 | |||
BC019952.1 | - | 4 | 388 | 4021772 |