Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028189.3 | NP_082465.3 | 2 | 294 | ||
GenBank mRNA | AK008674.1 | - | 333 | |||
AK144117.1 | - | 284 | ||||
AK144180.1 | - | 2 | 266 | |||
AY037785.1 | - | 263 | ||||
AY039028.1 | - | 1 | 213 | |||
BC026418.1 | - | 2 | 286 | 4218953 | ||
BY753510.1 | - | 2 | 294 |