Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fryl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028194.2 | NP_082470.2 | 44, 45 | 6201 | ||
GenBank mRNA | AB093265.1 | - | 7, 8 | 1025 | ||
BC051118.1 | - | 3, 4 | 795 |