Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172073.1 | NP_001165544.1 | 3, 4 | 466 | ||
NM_028209.2 | NP_082485.1 | 3, 4 | 465 | |||
GenBank mRNA | AK005923.1 | - | 3, 4 | 467 | ||
AK012652.1 | - | 3, 4 | 411 | |||
AK088684.1 | - | 3, 4 | 427 | |||
AK133931.1 | - | 3, 4 | 430 | |||
BC025435.1 | - | 3, 4 | 414 | 5004463 |