Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006304.1 | NP_006295.1 | 2 | 283 | ||
GenBank mRNA | AK094899.1 | - | 1 | 2557 | ||
AK291070.1 | - | 2 | 263 | |||
AK309241.1 | - | 2 | 241 | |||
BC032782.1 | - | 2 | 243 | 5166837 | ||
CR456887.1 | - | 2 | 155 | |||
U41515.1 | - | 2 | 283 |