Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163734.1 | NP_001157206.1 | 5, 6 | 782 | ||
NM_028244.2 | NP_082520.2 | 8, 9 | 1098 | |||
GenBank mRNA | AK011155.1 | - | 1, 2 | 24 | ||
AK129078.1 | - | 5, 6 | 782 | |||
BC016569.1 | - | 8, 9 | 1098 | 3157173 |