Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163734.1 | NP_001157206.1 | 4 | 572 | ||
NM_028244.2 | NP_082520.2 | 7 | 888 | |||
GenBank mRNA | AK129078.1 | - | 4 | 572 | ||
BC016569.1 | - | 7 | 888 | 3157173 |