Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naalad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028279.3 | NP_082555.2 | 19 | 2289 | ||
GenBank mRNA | AK012270.1 | - | 6 | 864 | ||
AK133611.1 | - | 19 | 2399 | |||
AK135653.1 | - | 21 | 2432 | |||
AK161457.1 | - | 22 | 2595 | |||
AY243507.1 | - | 19 | 2289 | |||
BC108995.1 | - | 19 | 2273 | 40047859 | ||
BC108996.1 | - | 19 | 2273 | 40047860 |