Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028315.2 | NP_082591.2 | 3, 4 | 676 | ||
GenBank mRNA | AK012840.1 | - | 3, 4 | 593 | ||
BC027357.1 | - | 3, 4 | 646 | 4952415 | ||
CN705423.1 | - | 2, 3 | 281 |