Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028315.2 | NP_082591.2 | 5 | 813 | ||
GenBank mRNA | AK012840.1 | - | 5 | 730 | ||
AK038313.1 | - | 5 | 683 | |||
AK129259.1 | - | 2 | 1101 | |||
BC027357.1 | - | 5 | 783 | 4952415 | ||
CN705423.1 | - | 4 | 418 |