Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028339.1 | NP_082615.1 | 4 | 432 | ||
GenBank mRNA | AK013150.1 | - | 4 | 432 | ||
AK030429.1 | - | 4 | 450 | |||
AK050567.1 | - | 4 | 431 | |||
AK137059.1 | - | 4 | 429 | |||
AK150511.1 | - | 4 | 439 | |||
AK153027.1 | - | 4 | 435 | |||
AK170468.1 | - | 4 | 450 | |||
BC017603.1 | - | 4 | 432 | 4503129 | ||
BC021533.1 | - | 4 | 453 | 5354337 |