Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271606.1 | NP_001258535.1 | 2 | 951 | ||
NM_006317.4 | NP_006308.3 | 2 | 1126 | |||
GenBank mRNA | AF039656.1 | - | 2 | 917 | ||
AK295995.1 | - | 3 | 892 | |||
BC000518.2 | - | 2 | 1041 | 2822874 |