Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccser2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027045.1 | NP_081321.1 | 6 | 2287 | ||
NM_028407.3 | NP_082683.2 | 4 | 459 | |||
GenBank mRNA | AK005931.1 | - | 4 | 459 | ||
AK129295.1 | - | 5 | 1859 | |||
AK147383.1 | - | 6 | 2282 | |||
AK172153.1 | - | 6 | 2261 | |||
BC048641.1 | - | 4 | 458 | |||
BC060947.1 | - | 4 | 458 | |||
BC111465.1 | - | 5 | 668 | |||
BC139090.1 | - | 5 | 2075 | 8862112 |