Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006324.2 | NP_006315.1 | 2 | 293 | ||
XM_011522814.1 | XP_011521116.1 | 2 | 316 | |||
XM_011522815.1 | XP_011521117.1 | 2 | 316 | |||
GenBank mRNA | AB009285.1 | - | 2 | 265 | ||
AK226031.1 | - | 2 | 310 | |||
AK313948.1 | - | 2 | 235 | |||
AW160432.1 | - | 2 | 297 | |||
BC000991.2 | - | 2 | 289 | 3449836 | ||
BT009819.1 | - | 2 | 156 | |||
CR542111.1 | - | 2 | 156 | |||
D85939.1 | - | 2 | 265 | |||
D86549.1 | - | 1 | 92 | |||
LC057679.1 | - | 2 | 156 | |||
LC057682.1 | - | 2 | 156 | |||
LC057683.1 | - | 2 | 156 |