Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164627.1 | NP_001158099.1 | 5 | 331 | ||
NM_001164628.1 | NP_001158100.1 | 4 | 323 | |||
NM_001205334.1 | NP_001192263.1 | 5 | 351 | |||
NM_028455.4 | NP_082731.2 | 4 | 343 | |||
GenBank mRNA | AK014171.1 | - | 4 | 294 | ||
AK040142.1 | - | 4 | 293 | |||
AK086816.1 | - | 4 | 297 | |||
AK088242.1 | - | 4 | 299 | |||
BC005563.1 | - | 3 | 286 | 3965919 | ||
BC010306.1 | - | 4 | 313 | |||
BY723460.1 | - | 4 | 317 |