Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006327.3 | NP_006318.1 | 7 | 730 | ||
NR_073029.1 | 8 | 854 | ||||
NR_073030.1 | 6 | 671 | ||||
GenBank mRNA | AF030162.1 | - | 7 | 682 | ||
AK223331.1 | - | 7 | 670 | |||
AK304883.1 | - | 6 | 554 | |||
AY442341.1 | - | 7 | 682 | |||
BC062707.1 | - | 7 | 731 | 4772587 | ||
BC066951.1 | - | 7 | 680 | 5264814 | ||
BY799488.2 | - | 6 | 604 | |||
CD514420.1 | - | 8 | 730 |