Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122963.1 | NP_001116435.1 | 5 | 1722 | ||
NM_028487.4 | NP_082763.3 | 5 | 1722 | |||
GenBank mRNA | AK082704.1 | - | 5 | 1540 | ||
AK135728.1 | - | 5 | 1653 | |||
AK136894.1 | - | 5 | 1501 | |||
AK145318.1 | - | 5 | 1711 | |||
AK151283.1 | - | 4 | 943 | |||
AK161928.1 | - | 5 | 1510 | |||
AK163370.1 | - | 5 | 1570 | |||
AK163390.1 | - | 5 | 1724 | |||
AK169406.1 | - | 5 | 1517 | |||
AY382529.1 | - | 5 | 1520 | |||
BC067075.1 | - | 5 | 1496 | 30614988 | ||
BC076635.1 | - | 5 | 1552 | |||
BC094888.1 | - | 5 | 1515 | 30628044 |