Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122963.1 | NP_001116435.1 | 12 | 3036 | ||
NM_028487.4 | NP_082763.3 | 13 | 3096 | |||
GenBank mRNA | AK135728.1 | - | 13 | 3027 | ||
AK145318.1 | - | 12 | 3025 | |||
AK151283.1 | - | 11 | 2256 | |||
AK163370.1 | - | 13 | 2944 | |||
AK163390.1 | - | 13 | 3098 | |||
AK169406.1 | - | 13 | 2891 | |||
AK211093.1 | - | 1 | 311 | |||
AY382529.1 | - | 13 | 2893 | |||
BC016083.1 | - | 11 | 1890 | 4501399 | ||
BC067075.1 | - | 12 | 2810 | 30614988 | ||
BC076635.1 | - | 12 | 2866 | |||
BC094888.1 | - | 12 | 2829 | 30628044 | ||
BC096442.1 | - | 13 | 2911 |