Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198836.1 | NP_001185765.1 | 2 | 510 | ||
NM_006328.3 | NP_006319.1 | 2 | 510 | |||
GenBank mRNA | AB209007.1 | - | 1 | 3210 | ||
AF080561.1 | - | 2 | 451 | |||
AF315632.1 | - | 2 | 466 | |||
AF315633.1 | - | 2 | 466 | |||
AK021768.1 | - | 2 | 462 | |||
AK222830.1 | - | 2 | 463 | |||
BC000488.2 | - | 2 | 446 | 2819856 | ||
BC023619.2 | - | 2 | 446 | |||
DB044990.1 | - | 2 | 511 | |||
KU178520.1 | - | 2 | 371 | |||
KU178521.1 | - | 2 | 371 |