Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276397.1 | NP_001263326.1 | 1 | 909 | ||
GenBank mRNA | AK005969.1 | - | 1 | 819 | ||
AK133385.1 | - | 1 | 801 | |||
BC050749.1 | - | 1 | 818 |