Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028604.2 | NP_082880.2 | 7, 8 | 775 | ||
GenBank mRNA | AF532979.1 | - | 7, 8 | 775 | ||
AK010722.1 | - | 7, 8 | 717 | |||
AK014181.1 | - | 6, 7 | 623 |